missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD105 (Endoglin) Mouse anti-Human, BB700, Clone: 266, BD Horizon™
Mouse Monoclonal Antibody
Marque: BD Biosciences 566529
Informations supplémentaires : Poids : 0.00810kg
Description
CD105 (Endoglin) Monoclonal antibody specifically detects CD105 (Endoglin) in Human samples. It is validated for Flow Cytometry.Spécification
CD105 (Endoglin) | |
Monoclonal | |
Brilliant Blue 700 (BB700) | |
25 Tests | |
Primary | |
Human | |
IgG1 κ |
Flow Cytometry | |
266 | |
Mouse | |
RUO | |
CD105 (Endoglin) | |
Refrigerate (2 to 8°C) |