missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD11c Mouse, V450, Clone: 3.9, enQuireBio™
Mouse Monoclonal Antibody
Marque: EnquireBio QAB24-V450-25Tests
Informations supplémentaires : Poids : 0.01000kg
Spécification
CD11c | |
Monoclonal | |
5μL/Test | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
ITGAX | |
Protein A/G Purified | |
RUO | |
3687 | |
IgG1 κ |
Flow Cytometry | |
3.9 | |
Violet Fluor 450 (V450) | |
XP_011544154 | |
Mouse | |
25 Tests | |
Primary | |
Store at 2-4°C. |