missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD142 Mouse anti-Human, BB700, Clone: HTF-1, BD Horizon™
Mouse Monoclonal Antibody
Marque: BD Biosciences 566562
Informations supplémentaires : Poids : 0.00810kg
Description
CD142 Monoclonal antibody specifically detects CD142 in Human samples. It is validated for Flow Cytometry.Spécification
CD142 | |
Monoclonal | |
Brilliant Blue 700 (BB700) | |
25 Tests | |
Primary | |
Refrigerate (2 to 8°C) | |
IgG1 κ |
Flow Cytometry | |
HTF-1 | |
Mouse | |
RUO | |
Human | |
Purified |