missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD152 Mouse anti-Human, Rhesus, Cynomolgus, Baboon, BB700, Clone: BNI3, BD Biosciences™
Mouse Monoclonal Antibody
Marque: BD Biosciences 566902
Informations supplémentaires : Poids : 0.00020kg
Description
CD152 Monoclonal antibody specifically detects CD152 in Human, Rhesus, Cynomolgus, Baboon samples. It is validated for Flow CytometrySpécification
CD152 | |
Monoclonal | |
Brilliant Blue 700 (BB700) | |
CTLA-4; AILIM; Cytotoxic T-lymphocyte protein 4 | |
Human CTLA4 Recombinant Protein | |
RUO | |
Human, Rhesus Monkey, Cynomolgus, Baboon | |
IgG2a κ |
Flow Cytometry | |
BNI3 | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
25 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed. |