missing translation for 'onlineSavingsMsg'
Learn More

CD172a (SIRP alpha) Monoclonal Antibody (P84), PE-Cyanine7, eBioscience™, Invitrogen™

Rat Monoclonal Antibody

Marque:  Invitrogen 25-1721-80

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 16360764

Cet article est actuellement indisponible ou discontinu. Veuillez consulter la page produit pour de possible alternative dans l’onglet
Voir les produits de remplacement »


Description

Description

Rat Monoclonal Antibody

CD172a (SIRP alpha) Monoclonal antibody specifically detects CD172a (SIRP alpha) in Mouse samples. It is validated for Flow Cytometry
Spécification

Spécification

CD172a (SIRP alpha)
Monoclonal
0.2 mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
P97797
SIRPA
Affinity chromatography
RUO
19261
4° C, store in dark, DO NOT FREEZE!
Liquid
Flow Cytometry
P84
PE-Cyanine7
SIRPA
2700084A17Rik; AI835480; AW536184; Bit; BOS_13304; BPTP3; Brain Ig-like molecule with tyrosine-based activation motifs; CD172 alpha; CD172 antigen-like family member A; CD172A; CFC; Inhibitory receptor SHPS-1; JMML; Macrophage fusion receptor; Macrophage membrane protein MFP150; METCDS; MFR; Myd1; MYD-1; MyD-1 antigen; NS1; P84; Protein-tyrosine phosphatase 1D; Protein-tyrosine phosphatase 2C; Protein-tyrosine phosphatase SYP; PTP1D; PTP-1D; PTP2C; PTP-2C; Ptpn11; PTPNS1; SAP-2; SHP substrate 1; SHP-1; SHP2; SHP-2; SHPS1; SHPS-1; SHPTP2; SH-PTP2; SH-PTP3; Signal-regulatory protein alpha-1; Signal-regulatory protein alpha-2; Signal-regulatory protein alpha-3; SIRP; Sirpa; SIRPalpha; Sirp-alpha-1; SIRPalpha2; Sirp-alpha-2; Sirp-alpha-3; swc3; Syp; Tyrosine-protein phosphatase non-receptor type 11; Tyrosine-protein phosphatase non-receptor type substrate 1
Rat
25 μg
Primary
Mouse
Antibody
IgG1 κ
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions