missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD1d Rat anti-Mouse, BB515, Clone: 1B1, BD Horizon™
Rat Monoclonal Antibody
Marque: BD Biosciences 565955
Informations supplémentaires : Poids : 0.01000kg
Description
CD1d Monoclonal antibody specifically detects CD1d in Mouse samples. It is validated for Flow cytometry.
CD1d Monoclonal antibody specifically detects CD1d in Mouse samples. It is validated for Flow cytometry.Spécification
CD1d | |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Rat | |
0.1 mg | |
Cell Biology | |
Mouse | |
Purified |
Flow Cytometry | |
1B1 | |
Brilliant Blue 515 (BB515) | |
Cd1d1; Cd1.1; Cd1a; Cd1d; Ly-38 | |
Mouse Cd1.1 cDNA-transfected RMA-S mouse T lymphoma and L929 cells | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BB515 under optimum conditions and unconjugated antibody was removed. | |
IgG2a κ |
For Research Use Only.