missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD2 Mouse, PerCP, Clone: RPA-2.10, enQuireBio™
Mouse Monoclonal Antibody
Marque: EnquireBio QAB1-PCP-25Tests
Informations supplémentaires : Poids : 0.01000kg
Spécification
CD2 | |
Monoclonal | |
5μL/Test | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
CD2 | |
Protein A/G Purified | |
RUO | |
914 | |
IgG1 κ |
Flow Cytometry | |
RPA-2.10 | |
PerCP | |
NP_001758 | |
Mouse | |
25 Tests | |
Primary | |
Store at 2-4°C. |