Learn More
CD27 Mouse anti-Human, PE, Clone: M-T271, BD
Mouse Monoclonal Antibody
Marque: BD Biosciences 555441
Informations supplémentaires : Poids : 0.02000kg
Description
The M-T271 monoclonal antibody specifically binds to CD27. CD27 presents asa type I transmembrane, disulphide-linked 110 kDa homodimer comprised of two polypeptide chains. The CD27 molecule is a lymphocyte-specific member of the TNF/NGF-R family, and is expressed on a subset of human thymocytes and on the majority of mature T lymphocytes, activated B cells and NK cells. CD27 is highly induced on T cells after TCR stimulation. CD27 binds to CD70 (also known as, CD27 ligand or CD27L) and may be involved in cellular interaction of T and B lymphocytes.
Spécification
CD27 | |
Monoclonal | |
PE | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
M-T271 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Human T-CLL cells | |
100 Tests | |
Immunology | |
Human | |
IgG1 κ |
For Research Use Only.