missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD274 / PD-L1 Rat, PE, Clone: 10F.9G2, enQuireBio™
Rat Monoclonal Antibody
Marque: EnquireBio QAB95-PE-25ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
CD274 / PD-L1 | |
Monoclonal | |
0.2mg/mL | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
Cd274 | |
Protein A/G Purified | |
RUO | |
60533 | |
IgG2b κ |
Flow Cytometry | |
10F.9G2 | |
PE | |
NP_068693 | |
Rat | |
25 μg | |
Primary | |
Store at 2-4°C. |