missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD3 Mouse anti-Human , BV786, Clone: OKT3, BD Biosciences™
Mouse Monoclonal Antibody
Marque: BD Biosciences 566781
Informations supplémentaires : Poids : 0.00060kg
Description
CD3 Monoclonal antibody specifically detects CD3 in Human samples. It is validated for Flow CytometrySpécification
CD3 | |
Monoclonal | |
Brilliant Violet 786 | |
CD3E; CD3e; T-cell surface antigen T3/Leu-4 epsilon; T3E; TCRE | |
Sheep Erythrocyte Rosette-purified Human T Cells | |
RUO | |
Human | |
IgG2a κ |
Flow Cytometry | |
OKT3 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Mouse | |
100 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BV786 under optimum conditions, and unconjugated antibody and free BD Horizon™ BV786 were removed. |