missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD304 Rat anti-Mouse , BV421, Clone: V46-1954, BD Biosciences™
Rat Monoclonal Antibody
Marque: BD Biosciences 567113
Informations supplémentaires : Poids : 0.00020kg
Description
CD304 Monoclonal antibody specifically detects CD304 in Mouse samples. It is validated for Flow CytometrySpécification
CD304 | |
Monoclonal | |
Brilliant Violet 421 | |
neuropilin-1; Nrp1; A5 protein; NP-1; NPN-1; Npn1; Nrp | |
Mouse CD304 Recombinant Protein | |
RUO | |
Mouse | |
IgG2a κ |
Flow Cytometry | |
V46-1954 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Rat | |
25 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed. |