missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD4 Mouse anti-Human, BUV563, Clone: SK3, BD Horizon™
Mouse Monoclonal Antibody
Marque: BD Biosciences 612913
Informations supplémentaires : Poids : 0.00020kg
Description
CD4 Monoclonal antibody specifically detects CD4 in Human samples. It is validated for Flow Cytometry.Spécification
CD4 | |
Monoclonal | |
Brilliant Ultraviolet 563 | |
25 Tests | |
Primary | |
Human | |
IgG1 κ |
Flow Cytometry | |
SK3 (also known as Leu3a) | |
Mouse | |
RUO | |
CD4 | |
Refrigerate (2 to 8°C) |