Learn More
CD56 (NCAM-1) Mouse, anti-Human, R718, Clone: B159, BD Horizon™
Mouse Monoclonal Antibody
Marque: BD Biosciences 566965
Informations supplémentaires : Poids : 0.00060kg
Description
The B159 monoclonal antibody specifically binds to CD56. CD56 is a heavily glycosylated adhesion protein that is present on a subpopulation of peripheral blood large granular lymphocytes that demonstrate natural killer activity. CD56 is also expressed on a subset of T cells but is not expressed on myeloid cells, erythrocytes or B cells. This antigen is a pan-NK-cell marker. CD56 is virtually identical to an isoform of the neural cell adhesion molecule (NCAM), a structure mediating homotypic and heterotypic cell-cell interactions.
Spécification
CD56 (NCAM-1) | |
Monoclonal | |
R718 | |
100 Tests | |
Primary | |
Refrigerate Upon Arrival |
Flow Cytometry | |
B159 | |
Mouse | |
RUO | |
Human | |
Liquid |