missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CEP63 Rabbit anti-Canine, Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 16268-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
This antibody can recognize all the four isoforms of CEP63 (56 kDa, 58 kDa, 63 kDa and 81 kDa).
CEP63 is a protein with six coiled-coil domains. The protein is localized to the centrosome, a non-membraneous organelle that functions as the major microtubule-organizing center in animal cells.Spécification
CEP63 | |
Polyclonal | |
Unconjugated | |
CEP63 | |
centrosomal protein 63kDa, Centrosomal protein of 63 kDa, CEP63 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
28135, 300963, 477075, 80254 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.59 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q3UPP8, Q4KLY0, Q96MT8 | |
Cep63 | |
CEP63 Fusion Protein Ag9335 | |
150 μL | |
Primary | |
Canine, Human, Mouse, Rat | |
Antibody | |
IgG |