missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHMP7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 16424-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
CHMP7 gene ontology annotations related to this gene include vacuolar transport.Spécification
CHMP7 | |
Polyclonal | |
Unconjugated | |
CHMP7 | |
CHMP family, member 7, CHMP7, Chromatin modifying protein 7 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
105513, 364419, 91782 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8R1T1, Q8WUX9 | |
Chmp7 | |
CHMP7 Fusion Protein Ag9625 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |