missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CITED2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 15021-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
There are two isoforms of CITED2 protein and the calculated molecular weight of them are 24kDa and 28 kDa.
The protein encoded by this gene inhibits transactivation of HIF1A-induced genes by competing with binding of HIF1a to p300-CH1. Mutations in this gene are a cause of cardiac septal defects. Three transcript variants encoding the same protein have been found for this gene.Spécification
CITED2 | |
Polyclonal | |
Unconjugated | |
CITED2 | |
CITED2, MRG 1, MRG1, MSG related protein 1, P35SRJ | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10370, 114490, 17684 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O35740, Q99967 | |
Cited2 | |
CITED2 Fusion Protein Ag7078 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |