Learn More
Claspin Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 23206-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
Xenopus claspin is an essential upstream regulator of checkpoint kinase 1 and triggers a checkpoint arrest of the cell cycle in the presence of DNA templates in Xenopus egg extracts. The human gene appears to be the homolog Xenopus claspin and its function has not been determined.Spécification
Claspin | |
Polyclonal | |
Unconjugated | |
CLSPN | |
CLASPIN, CLSPN, hClaspin | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
63967 | |
-20°C | |
Liquid |
Immunoprecipitation, Immunocytochemistry, Western Blot, Immunofluorescence | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9HAW4 | |
CLSPN | |
Claspin Fusion Protein Ag19637 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |