missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CMBL Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 17409-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
CMBL is a cysteine hydrolase of the dienelactone hydrolase family that is highly expressed in liver cytosol. CMBL preferentially cleaves cyclic esters, and it activates medoxomil-ester prodrugs in which the medoxomil moiety is linked to an oxygen atom.Spécification
CMBL | |
Polyclonal | |
Unconjugated | |
CMBL | |
CMBL | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
134147, 310201, 69574 | |
-20°C | |
Liquid |
Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q7TP52, Q8R1G2, Q96DG6 | |
Cmbl | |
CMBL Fusion Protein Ag11442 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |