missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CNGA3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 21657-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
This antibody detects CNGA3 with an apparent molecular weight of 98 kDa which has been reported.
This gene encodes a member of the cyclic nucleotide-gated cation channel protein family which is required for normal vision and olfactory signal transduction. Mutations in this gene are associated with achromatopsia (rod monochromacy) and color blindness. Two alternatively spliced transcripts encoding different isoforms have been described.Spécification
CNGA3 | |
Polyclonal | |
Unconjugated | |
CNGA3 | |
ACHM2, CCNC1, CCNCa, CCNCalpha, CNCG3, CNG 3, CNG channel alpha 3, CNG3, CNGA3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
1261, 12790 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q16281, Q9JJZ8 | |
CNGA3 | |
CNGA3 Fusion Protein Ag16385 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |