missing translation for 'onlineSavingsMsg'
Learn More

CNGA3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 21657-1-AP-20UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16831275

  • 140.00€ / 20µL
Expédition estimée: 30-05-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

This antibody detects CNGA3 with an apparent molecular weight of 98 kDa which has been reported.

This gene encodes a member of the cyclic nucleotide-gated cation channel protein family which is required for normal vision and olfactory signal transduction. Mutations in this gene are associated with achromatopsia (rod monochromacy) and color blindness. Two alternatively spliced transcripts encoding different isoforms have been described.
Spécification

Spécification

CNGA3
Polyclonal
Unconjugated
CNGA3
ACHM2, CCNC1, CCNCa, CCNCalpha, CNCG3, CNG 3, CNG channel alpha 3, CNG3, CNGA3
Rabbit
Antigen Affinity Chromatography
RUO
1261, 12790
-20°C
Liquid
Immunoprecipitation, Western Blot
0.23 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q16281, Q9JJZ8
CNGA3
CNGA3 Fusion Protein Ag16385
20 μL
Primary
Mouse, Human
Antibody
IgG
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions