missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CPNE6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 13782-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
This gene encodes a brain-specific member of the copine family, which is composed of calcium-dependent membrane-binding proteins. The gene product contains two N-terminal C2 domains, and one von Willebrand factor A domain. It may have a role in synaptic plasticity.Spécification
CPNE6 | |
Polyclonal | |
Unconjugated | |
CPNE6 | |
Copine 6, Copine VI, copine VI (neuronal), CPNE6, N COPINE, Neuronal copine | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
12891, 691478, 9362 | |
-20°C | |
Liquid |
Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O95741, Q9Z140 | |
CPNE6 | |
CPNE6 Fusion Protein Ag4789 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |