missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CXCL8/IL8 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 27095-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
Interleukin-8 (IL-8) is a chemokine produced by macrophages and other cell types such as epithelial cells. It is also synthesized by endothelial cells, which store IL-8 in their storage vesicles, the Weibel-Palade bodies. In humans, the interleukin-8 protein is encoded by the IL8 gene. Interleukin-8 (IL8) is a member of the CXC chemokine family. IL-8 is believed to play a role in the pathogenesis of bronchiolitis, a common respiratory tract disease caused by viral infection. This gene and other ten members of the CXC chemokine gene family form a chemokine gene cluster in a region mapped to chromosome 4q. The genes for IL8 have been co-localized on a 335-kb genomic fragment.Spécification
CXCL8/IL8 | |
Polyclonal | |
Unconjugated | |
CXCL8 | |
C X C motif chemokine 8, CXCL8, CXCL8,IL8, Emoctakin, GCP 1, GCP1, IL 8, IL 8(1 77), IL8, IL8/NAP1 form I Interleukin 8, IL8/NAP1 form II, IL8/NAP1 form VI, interleukin 8, MDNCF, MDNCF b IL 8(5 77) IL 8(6 77), NAF, NAF IL 8(7 77), NAP 1, NAP1, Neutrophil activating factor, Protein 3 10C | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
3576 | |
-20°C | |
Liquid |
Western Blot | |
0.4 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P10145 | |
CXCL8 | |
CXCL8/IL8 Fusion Protein Ag25640 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |