missing translation for 'onlineSavingsMsg'
Learn More

Cytokeratin 15 Mouse anti-Human, Mouse, Clone: 6B4F8, Proteintech

Mouse Monoclonal Antibody

Marque:  Proteintech 60247-1-IG-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16837285

  • 399.00€ / 150µL
Expédition estimée: 15-04-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Cytokeratin 15 is a member of the keratin gene family. The keratins are intermediate filament proteins responsible for the structural integrity of epithelial cells and are subdivided into cytokeratins and hair keratins. Most of the type I cytokeratins consist of acidic proteins which are arranged in pairs of heterotypic keratin chains and are clustered in a region on chromosome 17q21.2.
Spécification

Spécification

Cytokeratin 15
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P19012, Q61414
KRT15
Cytokeratin 15 Fusion Protein Ag0185
150 μL
Primary
Human, Mouse
Antibody
IgG1
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
6B4F8
Unconjugated
KRT15
CK 15, CK15, Cytokeratin 15, K15, K1CO, keratin 15, KRT15, KRTB
Mouse
Protein G
RUO
16665, 3866
-20°C
Liquid
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions