missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 8 Rabbit anti-Human, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 17514-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
This antibody is specifically against KRT8.
Cytokeratin 8 is a member of the type II keratin family clustered on the long arm of chromosome 12. Type I and type II keratins heteropolymerize to form intermediate-sized filaments in the cytoplasm of epithelial cells. Cutokeratin 8 typically dimerizes with keratin 18 to form an intermediate filament in simple single-layered epithelial cells. This protein plays a role in maintaining cellular structural integrity and also functions in signal transduction and cellular differentiation. Mutations in this gene cause cryptogenic cirrhosis.Spécification
Cytokeratin 8 | |
Polyclonal | |
Unconjugated | |
KRT8 | |
CARD2, CK 8, CK8, CYK8, Cytokeratin 8, K2C8, K8, keratin 8, KO, KRT8, Type II keratin Kb8 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
25626, 3856 | |
-20°C | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P05787, Q10758 | |
KRT8 | |
Peptide | |
150 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |