missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D2HGDH Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 13895-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
This gene encodes D-2hydroxyglutarate dehydrogenase, a mitochondrial enzyme belonging to the FAD-binding oxidoreductase/transferase type 4 family. This enzyme, which is most active in liver and kidney but also active in heart and brain, converts D-2-hydroxyglutarate to 2-ketoglutarate. Mutations in this gene are present in D-2-hydroxyglutaric aciduria, a rare recessive neurometabolic disorder causing developmental delay, epilepsy, hypotonia, and dysmorphic features.Spécification
D2HGDH | |
Polyclonal | |
Unconjugated | |
D2HGDH | |
D2HGD, D2HGDH | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
301624, 728294, 98314 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P84850, Q8CIM3, Q8N465 | |
D2HGDH | |
D2HGDH Fusion Protein Ag4857 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |