missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DARS2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 13807-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
The protein encoded by this gene belongs to the class-II aminoacyl-tRNA synthetase family. It is a mitochondrial enzyme that specifically aminoacylates aspartyl-tRNA. Mutations in this gene are associated with leukoencephalopathy with brainstem and spinal cord involvement and lactate elevation (LBSL).Spécification
DARS2 | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q3KRD0, Q6PI48, Q8BIP0 | |
Aspartate tRNA ligase, ASPRS, DARS2, LBSL, MT ASPRS, RP3 383J4.2 | |
Rabbit | |
IgG | |
150 μL | |
-20°C | |
Primary | |
226539, 304919, 55157 |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
DARS2 | |
Liquid | |
Dars2 | |
DARS2 Fusion Protein Ag4809 | |
Antigen Affinity Chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Human, Mouse, Rat |