missing translation for 'onlineSavingsMsg'
Learn More

DPH2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 12367-1-AP-20UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16830054

  • 121.38€ / 20µL
Expédition estimée: 30-09-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

This antibody works well in WB, IP, IHC and IF application.

This gene is one of two human genes similar to the yeast gene dph2. The yeast gene was identified by its ability to complement a diphthamide mutant strain, and thus probably functions in diphthamide biosynthesis. Diphthamide is a post-translationally modified histidine residue present in elongation factor 2 (EF2) that is the target of diphtheria toxin ADP-ribosylation. Two transcript variants encoding different isoforms have been found for this gene.
Spécification

Spécification

DPH2
Polyclonal
Unconjugated
DPH2
DPH2, DPH2 homolog, DPH2 homolog (S. cerevisiae), DPH2 like 2, DPH2L2, HsDph2
Rabbit
Antigen Affinity Chromatography
RUO
1802, 67728
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunocytochemistry, Immunoprecipitation, Immunofluorescence, Western Blot
0.17 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9BQC3, Q9CR25
DPH2
DPH2 Fusion Protein Ag3042
20 μL
Primary
Mouse, Human
Antibody
IgG
Vidéos
FDS
Documents
Certificats
Promotions

Promotions