Learn More
DPY30 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 16281-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
The DPY30 antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human DPY30. This antibody recognizes human, mouse, rat antigen. The DPY30 antibody has been validated for the following applications: IF, IHC,ELISA analysis.
Spécification
DPY30 | |
Polyclonal | |
Unconjugated | |
150 μL | |
84661 | |
Liquid |
Immunohistochemistry, Immunofluorescence, ELISA | |
400 μg/mL | |
Rabbit | |
Primary | |
Human, Mouse, Rat | |
IgG |