missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DULLARD Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 22088-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
Serine/threonine protein phosphatase forming with CNEP1R1 an active phosphatase complex that dephosphorylates and may activate LPIN1 and LPIN2. LPIN1 and LPIN2 are phosphatidate phosphatases that catalyze the conversion of phosphatidic acid to diacylglycerol and control the metabolism of fatty acids at different levels. May indirectly modulate the lipid composition of nuclear and/or endoplasmic reticulum membranes and be required for proper nuclear membrane morphology and/or dynamics. May also indirectly regulate the production of lipid droplets and triacylglycerol. May antagonize BMP signaling.Spécification
DULLARD | |
Polyclonal | |
Unconjugated | |
CTDNEP1 | |
DULLARD, HSA011916, NET56 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23399, 67181 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O95476, Q3TP92 | |
Ctdnep1 | |
DULLARD Fusion Protein Ag17288 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |