Learn More
EME1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque:  Proteintech 12975-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Code produit. 16830964
  • 121.38€ / 20µL
Expédition estimée: 28-09-2022
Description
This gene encodes a protein that complexes with methyl methanesulfonate-sensitive UV-sensitive 81 protein to form an endonuclease complex. The encoded protein interacts with specifc DNA structures including nicked Holliday junctions, 3′-flap structures and aberrant replication fork structures. This protein may be involved in repairing DNA damage and in maintaining genomic stability. Alternative splicing results in multiple transcript variants.
Spécification
Spécification
EME1
Polyclonal
Unconjugated
EME1
EME1, hMMS4, MMS4, MMS4 homolog, MMS4L
Rabbit
Antigen Affinity Chromatography
RUO
146956, 268465, 287634
-20°C
Liquid
Western Blot
0.25 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8BJW7, Q96AY2
Eme1
EME1 Fusion Protein Ag3580
20 μL
Primary
Human, Rat, Mouse