Learn More
ERG Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 14356-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
Transcriptional regulator. May participate in transcriptional regulation through the recruitment of SETDB1 histone methyltransferase and subsequent modification of local chromatin structure.Spécification
ERG | |
Polyclonal | |
Unconjugated | |
ERG | |
ERG, erg 3, p55, Transcriptional regulator ERG, Transforming protein ERG | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
13876, 170909, 2078 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P11308, P81270 | |
Erg | |
ERG Fusion Protein Ag5708 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |