Learn More
ERG Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque:  Proteintech 14356-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Code produit. 16832964
  • 140.00€ / 20µL
Expédition estimée: 31-05-2024
Description
Transcriptional regulator. May participate in transcriptional regulation through the recruitment of SETDB1 histone methyltransferase and subsequent modification of local chromatin structure.
Spécification
Spécification
ERG
Polyclonal
Unconjugated
ERG
ERG, erg 3, p55, Transcriptional regulator ERG, Transforming protein ERG
Rabbit
Antigen Affinity Chromatography
RUO
13876, 170909, 2078
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Western Blot, Immunoprecipitation
0.27 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P11308, P81270
Erg
ERG Fusion Protein Ag5708
20 μL
Primary
Human, Rat, Mouse