Learn More
F4/80 Rabbit anti-Mouse, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque:  Proteintech 28463-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Code produit. 16836155
  • 325.38€ / 150µL
Expédition estimée: 06-10-2022
Description
The F4/80 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of mouse F4/80. This antibody recognizes mouse antigen. The F4/80 antibody has been validated for the following applications: IF, IHC, ELISA analysis.
Spécification
Spécification
F4/80
Polyclonal
Unconjugated
150 μL
13733
IgG
Flow Cytometry, Immunofluorescence, Immunohistochemistry, Western Blot
367 μg/mL