Learn More
FAM83A Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 20618-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
The FAM83A antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human FAM83A. This antibody recognizes human antigen. The FAM83A antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Spécification
FAM83A | |
Polyclonal | |
Unconjugated | |
20 μL | |
84985 | |
IgG |
Western Blot, Immunohistochemistry, Immunofluorescence | |
333 μg/mL | |
Rabbit | |
Primary | |
Human |