missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FEM1C Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 14037-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
This antibody specifically recognizes the 69 kDa FEM1C protein.
Probable component of an E3 ubiquitin-protein ligase complex, in which it may act as a substrate recognition subunit.Spécification
FEM1C | |
Polyclonal | |
Unconjugated | |
FEM1C | |
EUROIMAGE686608, EUROIMAGE783647, fem 1 homolog c (C. elegans), FEM1 gamma, FEM1A, FEM1C, KIAA1785, Protein fem 1 homolog C | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
240263, 56929 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8CEF1, Q96JP0 | |
FEM1C | |
FEM1C Fusion Protein Ag5099 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |