missing translation for 'onlineSavingsMsg'
Learn More

FGD5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 20910-1-AP-20UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16830495

  • 140.00€ / 20µL
Expédition estimée: 12-04-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

FGD5 activates CDC42, a member of the Ras-like family of Rho- and Rac proteins, by exchanging bound GDP for free GTP. It mediates VEGF-induced CDC42 activation. It may regulate proangiogenic action of VEGF in vascular endothelial cells, including network formation, directional movement and proliferation and may play a role in regulating the actin cytoskeleton and cell shape.
Spécification

Spécification

FGD5
Polyclonal
Unconjugated
FGD5
FGD5, ZFYVE23
Rabbit
Antigen Affinity Chromatography
RUO
152273, 232237, 362402
-20°C
Liquid
Immunocytochemistry, Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin)
0.17 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q6ZNL6, Q80UZ0
Fgd5
FGD5 Fusion Protein Ag15037
20 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions