missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FGFBP1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 25006-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
This gene encodes a secreted fibroblast growth factor carrier protein. The encoded protein plays a critical role in cell proliferation, differentiation and migration by binding to fibroblast growth factors and potentiating their biological effects on target cells. The encoded protein may also play a role in tumor growth as an angiogenic switch molecule, and expression of this gene has been associated with several types of cancer including pancreatic and colorectal adenocarcinoma. A pseudogene of this gene is also located on the short arm of chromosome 4.Spécification
FGFBP1 | |
Polyclonal | |
Unconjugated | |
FGFBP1 | |
17 kDa HBGF binding protein, FGF binding protein 1, FGF BP, FGF BP1, FGFBP, FGFBP 1, FGFBP1, HBP17, Heparin-binding protein 17 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
9982 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.4 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q14512 | |
FGFBP1 | |
FGFBP1 Fusion Protein Ag6357 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |