missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FMC63 scFv, Rabbit anti-Mouse, Polyclonal Antibody, ACROBiosystems
Rabbit Polyclonal Antibody
Marque: ACROBiosystems FM3-S93-1mg
Informations supplémentaires : Poids : 0.00010kg
Description
Recognizes FMC63 scFv specifically, (recommended for ADA assay) (MALS verified)Spécification
FMC63 | |
Polyclonal | |
Lyophilized from 0.22μm filtered solution in PBS, pH7.4. Normally trehalose isadded as protectant before lyophilization. | |
1 mg | |
Primary | |
Mouse | |
Anti-idiotypic Antibodies |
ELISA | |
Unconjugated | |
Rabbit | |
RUO | |
Recognizes FMC63 scFv specifically | |
-20°C | |
Powder |