missing translation for 'onlineSavingsMsg'
Learn More

FXYD3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 15853-1-AP-20UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16815234

  • 140.00€ / 20µL
Expédition estimée: 30-05-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

FXYD3 belongs to a small family of FXYD-domain containing regulators of Na+/K+ ATPases which share a 35-amino acid signature sequence domain, beginning with the sequence PFXYD, and containing 7 invariant and 6 highly conserved amino acids. This gene encodes a cell membrane protein that may regulate the function of ion-pumps and ion-channels. This gene may also play a role in tumor progression. Alternative splicing results in multiple transcript variants encoding distinct isoforms.
Spécification

Spécification

FXYD3
Polyclonal
Unconjugated
FXYD3
FXYD3, Mammary tumor 8 kDa protein, MAT 8, MAT8, Phospholemman like, PLML
Rabbit
Antigen Affinity Chromatography
RUO
116831, 17178, 5349
-20°C
Liquid
Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
0.19 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P59645, Q14802, Q61835
Fxyd3
FXYD3 Fusion Protein Ag8672
20 μL
Primary
Human, Rat, Mouse
Antibody
IgG
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions