missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GDPD2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 25074-1-AP-150UL
Code nomenclature Nacres: NA.41
Informations supplémentaires : Poids : 0.00010kg
Description
Has glycerophosphoinositol inositolphosphodiesterase activity and specifically hydrolyzes glycerophosphoinositol, with no activity for other substrates such as glycerophosphoinositol 4-phosphate, glycerophosphocholine, glycerophosphoethanolamine, and glycerophosphoserine. Accelerates the program of osteoblast differentiation and growth. May play a role in remodeling of the actin cytoskeleton.Spécification
GDPD2 | |
Polyclonal | |
Unconjugated | |
GDPD2 | |
GDE3, GDPD2, OBDPF | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
54857, 71584 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.31 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9ESM6, Q9HCC8 | |
GDPD2 | |
GDPD2 Fusion Protein Ag18377 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |