Learn More
GIT1 Rabbit anti-Human, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque:  Proteintech 26247-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Code produit. 16834765
  • 399.00€ / 150µL
Expédition estimée: 30-05-2024
Description
N-myristoyltransferase (NMT) catalyzes the reaction of N-terminal myristoylation of many signaling proteins. It transfers myristic acid from myristoyl coenzyme A to the amino group of a protein′s N-terminal glycine residue. Biochemical evidence indicates the presence of several distinct NMTs, varying in apparent molecular weight and /or subcellular distribution. The predicted 498-amino acid of human NMT2 protein shares 77% and 96% sequence identity with human NMT1 and mouse Nmt2 comprise two distinct families of N-myristoyltransferases.
Spécification
Spécification
GIT1
Polyclonal
Unconjugated
GIT1
ARF GAP GIT1, CAT 1, CAT1, GIT1, GRK interacting protein 1
Rabbit
Antigen Affinity Chromatography
RUO
28964, 83709
-20°C
Liquid
Western Blot
0.23 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9Y2X7, Q9Z272
GIT1
GIT1 Fusion Protein Ag24648
150 μL
Primary
Human, Rat