missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GLO1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 15140-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
The enzyme encoded by this gene is responsible for the catalysis and formation of S-lactoyl-glutathione from methylglyoxal condensation and reduced glutathione. Glyoxalase I is linked to HLA and is localized to 6p21.3-p21.1, between HLA and the centromere.Spécification
GLO1 | |
Polyclonal | |
Unconjugated | |
GLO1 | |
Aldoketomutase, GLO1, GLOD1, Glx I, GLYI, glyoxalase I, Ketone aldehyde mutase, Lactoylglutathione lyase, Methylglyoxalase | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
109801, 2739, 294320 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
1.2 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q04760, Q6P7Q4, Q9CPU0 | |
Glo1 | |
GLO1 Fusion Protein Ag7314 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |