missing translation for 'onlineSavingsMsg'
Learn More

GLYR1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 14833-1-AP-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16893684

  • 349.00€ / 150µL
Expédition estimée: 09-12-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

GLYR1 may have oxidoreductase activity. It regulates p38 MAP kinase activity by mediating stress activation of p38alpha/MAPK14 and specifically regulating MAPK14 signaling. It indirectly promotes phosphorylation of MAPK14 and activation of ATF2. The phosphorylation of MAPK14 requires upstream activity of MAP2K4 and MAP2K6. It is recruited on chromatin, recognizes and binds trimethylated ′Lys-36′ of histone H3 (H3K36me3).
Spécification

Spécification

GLYR1
Polyclonal
Unconjugated
GLYR1
BM045, Glyoxylate reductase 1 homolog, GLYR1, HIBDL, N PAC, N-PAC, Nuclear protein NP60, Nuclear protein of 60 kDa, Putative oxidoreductase GLYR1
Rabbit
Antigen Affinity Chromatography
RUO
360477, 74022, 84656
-20°C
Liquid
Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry
0.35 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q49A26, Q5RKH0, Q922P9
Glyr1
GLYR1 Fusion Protein Ag6637
150 μL
Primary
Rat, Mouse, Human
Antibody
IgG
Vidéos
FDS
Documents
Certificats
Promotions

Promotions