missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GNAI2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 11136-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
The protein encoded by this gene is an alpha subunit of guanine nucleotide binding proteins (G proteins). The encoded protein contains the guanine nucleotide binding site and is involved in the hormonal regulation of adenylate cyclase. Several transcript variants encoding different isoforms have been detected for this gene, but the full-length nature of only two are known so far.Spécification
GNAI2 | |
Polyclonal | |
Unconjugated | |
GNAI2 | |
GIP, GNAI2, GNAI2B, H_LUCA15.1, H_LUCA16.1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
14678, 2771, 81664 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation, Immunocytochemistry, Immunofluorescence | |
0.32 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P04897, P04899, P08752 | |
Gnai2 | |
GNAI2 Fusion Protein Ag1597 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |