missing translation for 'onlineSavingsMsg'
Learn More

Grancalcin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 15787-1-AP-20UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16835124

  • 121.38€ / 20µL
Expédition estimée: 28-09-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

This gene product, grancalcin, is a calcium-binding protein abundant in neutrophils and macrophages. It belongs to the penta-EF-hand subfamily of proteins which includes sorcin, calpain, and ALG-2. Grancalcin localization is dependent upon calcium and magnesium. In the absence of divalent cation, grancalcin localizes to the cytosolic fraction; with magnesium alone, it partitions with the granule fraction; and in the presence of magnesium and calcium, it associates with both the granule and membrane fractions, suggesting a role for grancalcin in granule-membrane fusion and degranulation.
Spécification

Spécification

Grancalcin
Polyclonal
Unconjugated
GCA
GCA, GCL, Grancalcin
Rabbit
Antigen Affinity Chromatography
RUO
227960, 25801, 295647
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.25 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P28676, Q8VC88
Gca
Grancalcin Fusion Protein Ag8486
20 μL
Primary
Mouse, Human, Rat
Antibody
IgG
Vidéos
FDS
Documents
Certificats
Promotions

Promotions