Learn More
GRN1/G Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 25823-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
The GRN1/G antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human GRN1/G. This antibody recognizes human antigen. The GRN1/G antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Spécification
GRN1/G | |
Polyclonal | |
Unconjugated | |
150 μL | |
2896 | |
IgG |
Western Blot, Immunohistochemistry, ELISA | |
260 μg/mL | |
Rabbit | |
Primary | |
Human |