missing translation for 'onlineSavingsMsg'
Learn More

HECTD3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 11487-1-AP-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16828603

  • 399.00€ / 150µL
Expédition estimée: 15-04-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

The protein encoded by this gene transfers ubiquitin from an E2 ubiquitin-conjugating enzyme to targeted substrates, leading to the degradation of those substrates. The encoded protein has been shown to transfer ubiquitin to TRIOBP to facilitate cell cycle progression, and to STX8.
Spécification

Spécification

HECTD3
Polyclonal
Unconjugated
HECTD3
HECT domain containing 3, HECTD3, RP11 69J16.1
Rabbit
Antigen Affinity Chromatography
RUO
76608, 79654
-20°C
Liquid
Immunoprecipitation, Western Blot
0.27 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q3U487, Q5T447
Hectd3
HECTD3 Fusion Protein Ag1998
150 μL
Primary
Human, Mouse
Antibody
IgG
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions