Learn More
HIC2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 22788-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
HIC2 contains 5 C2H2-type zinc fingers and 1 BTB (POZ) domain. It belongs to the krueppel C2H2-type zinc-finger protein family, Hic subfamily and is a transcriptional repressor.Spécification
HIC2 | |
Polyclonal | |
Unconjugated | |
HIC2 | |
Hic 2, Hic 3, HIC2, HRG22, hypermethylated in cancer 2, KIAA1020, ZBTB30 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23119 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.61 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q96JB3 | |
HIC2 | |
HIC2 Fusion Protein Ag18789 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |