missing translation for 'onlineSavingsMsg'
Learn More
Learn More
His--Protein Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 10560-1-IG-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
His refers to histidine residues on the N- or C-terminus of a protein.Spécification
His-Bacteria-Protein | |
Polyclonal | |
Unconjugated | |
His Bacteria Protein | |
His-Bacteria-Protein Fusion Protein Ag0050 | |
20 μL | |
Primary | |
-20°C | |
Liquid |
Western Blot | |
0.77 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Rabbit | |
Protein A | |
RUO | |
Human | |
Antibody | |
IgG |