missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA-B7 Monoclonal Antibody (BB7.1), PE-Cyanine7, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Marque: Affymetrix eBioscience 25-5917-41
Informations supplémentaires : Poids : 0.01000kg
Description
Mouse Monoclonal Antibody
HLA-B7 Monoclonal antibody specifically detects HLA-B7 in Human samples. It is validated for Flow CytometrySpécification
HLA-B7 | |
Monoclonal | |
5 μL/Test | |
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
P01889 | |
HLA-B | |
Papain solubilized HLA-B7 | |
25 Tests | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
Flow Cytometry | |
BB7.1 | |
PE-Cyanine7 | |
HLA-B | |
AS; Aw-33; Aw-74; B-4901; B-5001; Bw-47; Bw-50; CDABP0067; DAQB-90C11.16; FLJ26655; HEL-S-83; HLA class I histocompatibility antigen, A-1 alpha chain; HLA class I histocompatibility antigen, A-3 alpha chain; HLA class I histocompatibility antigen, B-7 alpha chain; HLAA; HLA-A; HLA-A11; HLA-A33; HLAB; HLA-B; HLA-B 1502; HLA-B*45ZJ; HLA-B15; HLA-B-3506; HLA-B39; HLA-B-3905; HLA-B49; HLA-B50; HLA-B55; HLA-B-5502; HLA-B-5602; HLA-B59; HLA-B61; HLA-Cw; HLA-DQB1; HLA-DRB1; MHC 1; MHC class I antigen A*1; MHC class I antigen A*3; MHC class I antigen B*7; MHC1; SPDA1 | |
Mouse | |
Affinity chromatography | |
RUO | |
3106 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |